Acetoin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Acetyl methyl carbinol, 3-Hydroxybutanone, 3-Hydroxy-2-butanone

M-Clarity™ quality level = MQ200

8.20664.0100 8.20664.0250 8.20664.0100 8.20664.0250
EM8.20664.0100CS 2317.86 CAD
EM8.20664.0100 EM8.20664.0250 CA8.20664.0100 CA8.20664.0250
Acetoin for synthesis, Sigma-Aldrich®
Acetoin
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 148
Melting Pt: 20
MDL Number: MFCD00038696
CAS Number: 513-86-0
UN: 2621
ADR: 3 III
Merck Index: 13,00065

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 20 °C Above 20°C conversion of dimer to monomer (15°C)
Molar Mass 176.22 g/mol
Boiling Point 147 - 148 °C
Flash Point 45 °C
Refractive Index 1.42 (20 °C, 589 nm)
Density 1.012 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR