1-Hexyne for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20643.0025 8.20643.0100 8.20643.0025 8.20643.0100
EM8.20643.0025CS 7373.86 CAD
EM8.20643.0025 EM8.20643.0100 CA8.20643.0025 CA8.20643.0100
1-Hexyne for synthesis, Sigma-Aldrich®
1-Hexyne
Formula: C₆H₁₀
MDL Number: MFCD00009504
CAS Number: 693-02-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Solubility 0.36 g/l (25 °C) (experimental)
Melting Point -132 °C
Molar Mass 82.14 g/mol
Boiling Point 71 - 72 °C (1013 hPa)
Vapor Pressure 3535 kPa
Flash Point -20 °C
Refractive Index 1.3974 (20 °C, 589 nm)
Density 0.72 g/cm3 (20 °C)
Ignition Point 263 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR