N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DPPD, N,N′-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-1,4-phenylenediamine

M-Clarity™ quality level = MQ200

8.20536.0250 8.20536.9010 8.20536.0250 8.20536.9010
EM8.20536.0250CS 5379.9 CAD
EM8.20536.0250 EM8.20536.9010 CA8.20536.0250 CA8.20536.9010
N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by GC) for synthesis, Sigma-Aldrich®
N,N'-Diphenyl-p-phenylenediamine
Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Boiling Pt: 220…225 °C (1.33 hPa)
Melting Pt: 135 °C
Density: 1.3 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003015
CAS Number: 74-31-7

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 135 °C
Molar Mass 260.33 g/mol
Bulk Density 560 kg/m3
Boiling Point 220 - 225 °C (1.33 hPa)
Density 1.3 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR