5,5-Diphenylhydantoin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20534.0005 8.20534.0250 8.20534.0005 8.20534.0250
EM8.20534.0005CS 161510.76 CAD
EM8.20534.0005 EM8.20534.0250 CA8.20534.0005 CA8.20534.0250
5,5-Diphenylhydantoin for synthesis, Sigma-Aldrich®
5,5-Diphenylhydantoin
Formula: C₁₅H₁₂N₂O₂
MDL Number: MFCD00005264
CAS Number: 57-41-0

Order Now

Specification Test Results

Solubility 0.032 g/l (22 °C) (experimental)
Melting Point 292 - 295 °C
Molar Mass 252.27 g/mol
Bulk Density 400 - 450 kg/m3
Ignition Point 550 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR