3,4-Dihydroxybenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Protocatechuic aldehyde, Protocatechualdehyde

M-Clarity™ quality level = MQ200

8.20475.0025 8.20475.0100 8.20475.0025 8.20475.0100
EM8.20475.0025CS 37160.02 CAD
EM8.20475.0025 EM8.20475.0100 CA8.20475.0025 CA8.20475.0100
3,4-Dihydroxybenzaldehyde for synthesis, Sigma-Aldrich®
3,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 295
MDL Number: MFCD00003370
CAS Number: 139-85-5
Merck Index: 12,08079

Order Now

Specification Test Results

Solubility 6.3 g/l (25 °C)
Melting Point 152 - 156 °C
Molar Mass 138.12 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR