2,4-Dichlorobenzaldehyde ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20433.0100 8.20433.0500 8.20433.9025 8.20433.0100 8.20433.0500 8.20433.9025
EM8.20433.0100CS 7177.94 CAD
EM8.20433.0100 EM8.20433.0500 EM8.20433.9025 CA8.20433.0100 CA8.20433.0500 CA8.20433.9025
2,4-Dichlorobenzaldehyde ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
2,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00003305
CAS Number: 874-42-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 68 - 71 °C
Molar Mass 175.01 g/mol
Bulk Density 700 kg/m3
Boiling Point 233 °C (1013 hPa)
Flash Point 135 °C
Ignition Point 440 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR