Diethylamine hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20393.0005 8.20393.0250 8.20393.1000 8.20393.9025 8.20393.0005 8.20393.1000 8.20393.9025
EM8.20393.0005CS 50980.79 CAD
EM8.20393.0005 EM8.20393.0250 EM8.20393.1000 EM8.20393.9025 CA8.20393.0005 CA8.20393.0250 CA8.20393.1000 CA8.20393.9025
Diethylamine hydrochloride for synthesis, Sigma-Aldrich®
Diethylamine hydrochloride
Formula: C₄H₁₂ClN
Melting Pt: 231.8 °C
Density: 1.07 g/cm3 at 20.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00012499
CAS Number: 660-68-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 222 °C
Molar Mass 109.60 g/mol
Bulk Density 1100 kg/m3
Boiling Point 320 - 330 °C
pH 4.5 - 6.5 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR