n-Decane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20383.0250 8.20383.1000 8.20383.9037 8.20383.0250 8.20383.1000 8.20383.9037
EM8.20383.0250CS 910.08 CAD
EM8.20383.0250 EM8.20383.1000 EM8.20383.9037 CA8.20383.0250 CA8.20383.1000 CA8.20383.9037
n-Decane for synthesis, Sigma-Aldrich®
n-Decane
Formula: H₃C(CH₂)₈CH₃
Boiling Pt: 174 °C (1013 mbar)
Melting Pt: –30 °C
Density: 0.73 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
UN: 2247
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.00005 g/l (25 °C)
Melting Point -30 °C
Molar Mass 142.28 g/mol
Boiling Point 174 °C
Vapor Pressure 1 hPa (16 °C)
Flash Point 51 °C
Explosion Limit 0.7 - 5.4 %(V)
Density 0.73 g/cm3 (20 °C)
Ignition Point 208 °C
Viscosity Kinematic  - 7 mm2/s (40 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR