cis-1,2-Cyclohexanedicarboxylic anhydride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20365.0100
8.20365.1000
8.20365.0100
8.20365.1000
EM8.20365.0100CS
17258.34
CAD
EM8.20365.0100
EM8.20365.1000
CA8.20365.0100
CA8.20365.1000
cis-1,2-Cyclohexanedicarboxylic anhydride for synthesis, Sigma-Aldrich®
cis-1,2-Cyclohexanedicarboxylic anhydride
Formula:
C₈H₁₀O₃ |
MDL Number:
MFCD00005926 CAS Number: 13149-00-3 |
Specification Test Results
Solubility | 7 g/l (20 °C) (slow decomposition) |
Melting Point | 30 - 33 °C |
Molar Mass | 154.16 g/mol |
Boiling Point | 296 °C (1013 hPa) |
Vapor Pressure | 0.01 hPa (20 °C) |
Flash Point | 152 °C |
Density | 1.23 g/cm3 (20 °C) |
Ignition Point | 395 °C DIN 51794 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...