Cyanuric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

M-Clarity™ quality level = MQ200

8.20358.0005 8.20358.0250 8.20358.1000 8.20358.9050 8.20358.0005 8.20358.0250 8.20358.1000 8.20358.9050
EM8.20358.0005CS 42389.17 CAD
EM8.20358.0005 EM8.20358.0250 EM8.20358.1000 EM8.20358.9050 CA8.20358.0005 CA8.20358.0250 CA8.20358.1000 CA8.20358.9050
Cyanuric acid for synthesis, Sigma-Aldrich®
Cyanuric acid
Formula: C₃H₃N₃O₃
MW: 129.08 g/mol
Melting Pt: 320…330 °C (dec.)
Density: 2.5 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082990
CAS Number: 108-80-5
Merck Index: 13,02728

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point 320 - 330 °C (decomposition)
Molar Mass 129.07 g/mol
Bulk Density 550 kg/m3
Vapor Pressure  - 0.0001 Pa (25 °C)
Density 2.5 g/cm3 (20 °C)
pH 3.8 - 4.0 (H2O, 20 °C) (saturated solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR