Curcumin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20354.0010 8.20354.0050 8.20354.0010 8.20354.0050 8.20354.0002
EM8.20354.0010CS 9713.84 CAD
EM8.20354.0010 EM8.20354.0050 CA8.20354.0010 CA8.20354.0050 CA8.20354.0002
Curcumin for synthesis, Sigma-Aldrich®
Curcumin
Formula: C₂₁H₂₀O₆
Storage Temperature: Ambient
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5

Order Now

Specification Test Results

Assay (HPLC, area%) ≥ 75.0 % (a/a)
Bisdemethoxycurcumin (HPLC; Area%) ≤ 5.0 % (a/a)
Demethoxycurcumin (HPLC; Area%) ≤ 20.0 % (a/a)
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR