Chlorocyclohexane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20287.0250 8.20287.1000 8.20287.0250 8.20287.1000
EM8.20287.0250CS 943.26 CAD
EM8.20287.0250 EM8.20287.1000 CA8.20287.0250 CA8.20287.1000
Chlorocyclohexane for synthesis, Sigma-Aldrich®
Chlorocyclohexane
Formula: C₆H₁₁Cl
MDL Number: MFCD00003822
CAS Number: 542-18-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Solubility 0.40 g/l (20 °C)
Melting Point -44 °C
Molar Mass 118.6 g/mol
Boiling Point 141 - 143 °C (1013 hPa)
Vapor Pressure 7 hPa (20 °C)
Flash Point 34 °C
Refractive Index 1.4626 (20 °C, 589 nm)
Explosion Limit 1.1 - 7.5 %(V)
Density 1.00 g/cm3 (20 °C)
Ignition Point 290 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR