5-Isopropyl-2-methylphenol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Carvacrol, 5-Isopropyl-o-cresol, Isothymol, 5-Isopropyl-2-methylphenol
M-Clarity™ quality level = MQ200
8.20258.0050
8.20258.0250
8.20258.0050
8.20258.0250
EM8.20258.0050CS
16139.7
CAD
EM8.20258.0050
EM8.20258.0250
CA8.20258.0050
CA8.20258.0250
5-Isopropyl-2-methylphenol for synthesis, Sigma-Aldrich®
5-Isopropyl-2-methylphenol
Formula:
C₁₀H₁₄O MW: 150.22 g/mol |
MDL Number:
MFCD00002236 CAS Number: 499-75-2 UN: 3265 ADR: 8,III |
Specification Test Results
Solubility | 1.25 g/l (25 °C) |
Melting Point | 3 °C |
Molar Mass | 150.22 g/mol |
Boiling Point | 237 - 239 °C |
Flash Point |
|
Refractive Index | 1.5229 (20 °C, 589 nm) |
Density | 0.98 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...