4-tert-Butylbenzoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 4-(tert-Butyl)benzoic acid

M-Clarity™ quality level = MQ200

8.20238.0100 8.20238.1000 8.20238.0100 8.20238.1000
EM8.20238.0100CS 963.8 CAD
EM8.20238.0100 EM8.20238.1000 CA8.20238.0100 CA8.20238.1000
4-tert-Butylbenzoic acid for synthesis, Sigma-Aldrich®
4-tert-Butylbenzoic acid
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 283
Flash Pt: 180
MDL Number: MFCD00002563
CAS Number: 98-73-7
UN: 2811

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 165 - 167 °C
Molar Mass 178.23 g/mol
Bulk Density 340 kg/m3
Boiling Point 280 °C (decomposition)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 180 °C
Density 0.6 g/cm3 (20 °C)
pH 3.9 (H2O) (saturated solution)
Ignition Point 510 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR