4,4'-Bis(diethylamino)benzophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone

M-Clarity™ quality level = MQ200

8.20161.0050 8.20161.0250 8.20161.0050 8.20161.0250
EM8.20161.0050CS 504.02 CAD
EM8.20161.0050 EM8.20161.0250 CA8.20161.0050 CA8.20161.0250
4,4'-Bis(diethylamino)benzophenone for synthesis, Sigma-Aldrich®
4,4'-Bis(diethylamino)benzophenone
Formula: C₂₁H₂₈N₂O
MW: 324.47 g/mol
MDL Number: MFCD00009044
CAS Number: 90-93-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 90 - 95 °C
Molar Mass 324.46 g/mol
Bulk Density 400 kg/m3
Flash Point 151 °C
pH 4 - 6 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR