Dimethyl succinate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester
M-Clarity™ quality level = MQ200
8.20150.0250
8.20150.1000
8.20150.0250
8.20150.1000
EM8.20150.0250CS
970.12
CAD
EM8.20150.0250
EM8.20150.1000
CA8.20150.0250
CA8.20150.1000
Dimethyl succinate for synthesis, Sigma-Aldrich®
Dimethyl succinate
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 200 |
MDL Number:
MFCD00008466 CAS Number: 106-65-0 Merck Index: 12,09038 |
Specification Test Results
Solubility | 75 g/l (20 °C) |
Melting Point | 18 - 19 °C |
Molar Mass | 146.14 g/mol |
Boiling Point | 196 °C (1013 hPa) |
Vapor Pressure | 0.2 hPa (20 °C) |
Flash Point | 94 °C |
Refractive Index | 1.419 (20 °C, 589 nm) |
Explosion Limit | 1.0 - 8.5 %(V) |
Density | 1.12 g/cm3 (20 °C) |
Ignition Point | 365 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...