Saccharin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: o-Sulphobenzimide, o-Benzoic sulphimide, o-Benzoic sulfimide, o-Sulfobenzimide

M-Clarity™ quality level = MQ200

8.20128.0250 8.20128.1000 8.20128.5000 8.20128.0250 8.20128.1000 8.20128.5000
EM8.20128.0250CS 1835.96 CAD
EM8.20128.0250 EM8.20128.1000 EM8.20128.5000 CA8.20128.0250 CA8.20128.1000 CA8.20128.5000
Saccharin for synthesis, Sigma-Aldrich®
Saccharin
Formula: C₇H₅NO₃S
MDL Number: MFCD00005866
CAS Number: 81-07-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 228 - 230 °C
Molar Mass 183.18 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR