Ethanesulfonic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20016.0005 8.20016.0025 8.20016.0005 8.20016.0025
EM8.20016.0005CS 53535.14 CAD
EM8.20016.0005 EM8.20016.0025 CA8.20016.0005 CA8.20016.0025
Ethanesulfonic acid for synthesis, Sigma-Aldrich®
Ethanesulphonic acid
Formula: C₂H₆O₃S
MW: 110.13 g/mol
MDL Number: MFCD00007529
CAS Number: 594-45-6
UN: 2586
ADR: 8,III

Order Now

Specification Test Results

Solubility C2H6O3S
Melting Point C2H5SO3H
Molar Mass 110.13 g/mol
Boiling Point 136 - 140 °C (3 hPa)
Refractive Index 1.4335 (20 °C, 589 nm)
Density 1.34 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR