EDTA dipotassium salt dihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt

M-Clarity™ quality level = MQ200

8.19040.0025 8.19040.0100 8.19040.9025 8.19040.0025 8.19040.0100 8.19040.9025
EM8.19040.0025CS 5632.7 CAD
EM8.19040.0025 EM8.19040.0100 EM8.19040.9025 CA8.19040.0025 CA8.19040.0100 CA8.19040.9025
EDTA dipotassium salt dihydrate for synthesis, Sigma-Aldrich®
EDTA dipotassium salt dihydrate
Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
Melting Pt: 272 °C
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS Number: 25102-12-9
Merck Index: 13,03546

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 272 °C
Molar Mass 404.47 g/mol
Bulk Density 310 kg/m3
Vapor Pressure (20 °C)
pH 4 - 5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR