(±)-Mandelonitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.19016.0100 8.19016.0250 8.19016.0100 8.19016.0250
EM8.19016.0100CS 2461.64 CAD
EM8.19016.0100 EM8.19016.0250 CA8.19016.0100 CA8.19016.0250
(±)-Mandelonitrile for synthesis, Sigma-Aldrich®
(±)-Mandelonitrile
Formula: C₈H₇NO
MW: 133.15 g/mol
MDL Number: MFCD00004487
CAS Number: 532-28-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Solubility  - 1 g/l (20 °C)
Melting Point -10 °C
Molar Mass 133.15 g/mol
Boiling Point 170 °C (1013 hPa)
Flash Point 86 °C
Density 1.12 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR