Magnesium ethanolate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18894.0100 8.18894.0500 8.18894.0100 8.18894.0500
EM8.18894.0100CS 6590.18 CAD
EM8.18894.0100 EM8.18894.0500 CA8.18894.0100 CA8.18894.0500
Magnesium ethanolate for synthesis, Sigma-Aldrich®
Magnesium ethanolate
Formula: C₄H₁₀MgO₂
MDL Number: MFCD00009060
CAS Number: 2414-98-4
UN: 3205
ADR: 4.2,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 114.43 g/mol
Bulk Density 480 kg/m3
Ignition Point 70 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR