Ethoxytrimethylsilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18821.0025 8.18821.0100 8.18821.0025 8.18821.0100
EM8.18821.0025CS 5447.84 CAD
EM8.18821.0025 EM8.18821.0100 CA8.18821.0025 CA8.18821.0100
Ethoxytrimethylsilane for synthesis, Sigma-Aldrich®
Ethoxytrimethylsilane
Formula: C₅H₁₄OSi
Boiling Pt: 75…76 °C (1013 mbar)
Melting Pt: –83 °C
Density: 0.76 g/cm3 (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00009069
CAS Number: 1825-62-3
EINECS: 217-370-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Assay (GC, area%) ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.756 - 0.759
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR