Trimethyl orthoacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18804.0100 8.18804.0500 8.18804.0100 8.18804.0500
EM8.18804.0100CS 1546.82 CAD
EM8.18804.0100 EM8.18804.0500 CA8.18804.0100 CA8.18804.0500
Trimethyl orthoacetate for synthesis, Sigma-Aldrich®
Trimethyl orthoacetate
Formula: C₅H₁₂O₃
MDL Number: MFCD00008477
CAS Number: 1445-45-0
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 120.14 g/mol
Boiling Point 108 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Flash Point 15 °C
Refractive Index 1.3859 (25 °C, 589 nm)
Density 0.96 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR