Cyclopentane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Pentamethylene

M-Clarity™ quality level = MQ200

8.18769.0250 8.18769.1000 8.18769.0250 8.18769.1000
EM8.18769.0250CS 1540.5 CAD
EM8.18769.0250 EM8.18769.1000 CA8.18769.0250 CA8.18769.1000
Cyclopentane for synthesis, Sigma-Aldrich®
Cyclopentane
Formula: C₅H₁₀
MW: 70.13 g/mol
MDL Number: MFCD00001356
CAS Number: 287-92-3
UN: 1146
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dd230057521cc77c90840c7adfece9c74c0a3b1&vid=352021851b7701a448c5b11d43e94d925d03f802&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -93.3 °C
Molar Mass 70.13 g/mol
Boiling Point 49 °C
Vapor Pressure 346 hPa (20 °C)
Flash Point -42 °C
Refractive Index 1.4065 (20 °C)
Explosion Limit 1.5 - 8.7 %(V)
Density 0.75 g/cm3 (20 °C)
Ignition Point 380 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR