3-Chloro-4-fluoroaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18752.0025 8.18752.0100 8.18752.0025 8.18752.0100
EM8.18752.0025CS 14825.14 CAD
EM8.18752.0025 EM8.18752.0100 CA8.18752.0025 CA8.18752.0100
3-Chloro-4-fluoroaniline for synthesis, Sigma-Aldrich®
3-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
MDL Number: MFCD00007767
CAS Number: 367-21-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Melting Point 41 - 44 °C
Molar Mass 145.56 g/mol
Bulk Density 1000 kg/m3
Boiling Point 230 °C (1013 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 114 °C
Density 1.59 g/cm3
pH 7 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR