N,O-Bis(trimethylsilyl)acetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

M-Clarity™ quality level = MQ200

8.18727.0025 8.18727.0100 8.18727.0025 8.18727.0100
EM8.18727.0025CS 7972.68 CAD
EM8.18727.0025 EM8.18727.0100 CA8.18727.0025 CA8.18727.0100
N,O-Bis(trimethylsilyl)acetamide for synthesis, Sigma-Aldrich®
N,O-Bis(trimethylsilyl)acetamide
Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8 (3),II

Order Now

Specification Test Results

Solubility (20 °C) decomposes
Molar Mass 203.43 g/mol
Boiling Point 43 °C (11 hPa)
Vapor Pressure 10 hPa (50 °C)
Flash Point 40 °C
Density 0.83 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR