Urea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

M-Clarity™ quality level = MQ200

8.18710.1000 8.18710.5000 8.18710.1000 8.18710.5000
EM8.18710.1000CS 821.6 CAD
EM8.18710.1000 EM8.18710.5000 CA8.18710.1000 CA8.18710.5000
Urea for synthesis, Sigma-Aldrich®
Urea
Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
MDL Number: MFCD00008022
CAS Number: 57-13-6
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 590 g/l (20 °C)
Melting Point 133 °C
Molar Mass 60.06 g/mol
Bulk Density 720 - 760 kg/m3
Vapor Pressure  - 0.1 hPa (20 °C)
Density 1.34 g/cm3 (20 °C)
pH 9 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR