2,2-Dichloroacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18672.0100 8.18672.0500 8.18672.0100 8.18672.0500
EM8.18672.0100CS 10246.3 CAD
EM8.18672.0100 EM8.18672.0500 CA8.18672.0100 CA8.18672.0500
2,2-Dichloroacetamide for synthesis, Sigma-Aldrich®
2,2-Dichloroacetamide
Formula: C₂H₃Cl₂NO
MDL Number: MFCD00008015
CAS Number: 683-72-7
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 71 g/l (20 °C)
Melting Point 96 - 99 °C
Molar Mass 127.95 g/mol
Boiling Point 233 - 234 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR