2-Acrylamido-2-methylpropanesulfonic acid ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Acrylamido-2-methyl-1-propanesulphonic acid, 2-Acrylamido-2-methyl-1-propanesulfonic acid

M-Clarity™ quality level = MQ200

8.18667.0100 8.18667.0500 8.18667.9025 8.18667.0100 8.18667.0500 8.18667.9025
EM8.18667.0100CS 469.26 CAD
EM8.18667.0100 EM8.18667.0500 EM8.18667.9025 CA8.18667.0100 CA8.18667.0500 CA8.18667.9025
2-Acrylamido-2-methylpropanesulfonic acid ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
2-Acrylamido-2-methylpropanesulphonic acid
Formula: C₇H₁₃NO₄S
Melting Pt: 185 °C
Density: 1.1 g/cm3 at 15.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00007522
CAS Number: 15214-89-8

Order Now

Specification Test Results

Melting Point 190 °C
Molar Mass 207.25 g/mol
Flash Point 300 °C
Density 1.36 g/cm3 (20.0 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR