5-Methyl-3-heptanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Amyl ethyl ketone, Ethyl Amyl ketone

M-Clarity™ quality level = MQ200

8.18663.0100 8.18663.1000 8.18663.0100 8.18663.1000
EM8.18663.0100CS 1185 CAD
EM8.18663.0100 EM8.18663.1000 CA8.18663.0100 CA8.18663.1000
5-Methyl-3-heptanone for synthesis, Sigma-Aldrich®
5-Methyl-3-heptanone
Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 159
Melting Pt: -56.7
Flash Pt: 43
MDL Number: MFCD00009340
CAS Number: 541-85-5
UN: 2271
ADR: 3 III
Merck Index: 12,03810

Order Now

Specification Test Results

Solubility 3 g/l (20 °C)
Melting Point -57 °C
Molar Mass 128.22 g/mol
Boiling Point 157 - 162 °C (1013 hPa)
Vapor Pressure 2.7 hPa (25 °C)
Flash Point 43 °C
Refractive Index 1.4209 (20 °C, 589 nm)
Explosion Limit 0.9 - 48 %(V)
Density 0.82 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR