N-tert-Butylbenzylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine

M-Clarity™ quality level = MQ200

8.18660.0005 8.18660.0250 8.18660.0005 8.18660.0250
EM8.18660.0005CS 5063.9 CAD
EM8.18660.0005 EM8.18660.0250 CA8.18660.0005 CA8.18660.0250
N-tert-Butylbenzylamine for synthesis, Sigma-Aldrich®
N-tert-Butylbenzylamine
Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD00008798
CAS Number: 3378-72-1
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point -30 - -25 °C
Molar Mass 163.26 g/mol
Boiling Point 218 - 220 °C
Vapor Pressure 0.07 hPa (53 °C)
Flash Point 90 °C
Refractive Index 1.4968 (20 °C, 589 nm)
Density 0.90 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR