n-Undecane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Undecane

M-Clarity™ quality level = MQ200

8.18629.0100 8.18629.0500 8.18629.0100 8.18629.0500
EM8.18629.0100CS 1869.14 CAD
EM8.18629.0100 EM8.18629.0500 CA8.18629.0100 CA8.18629.0500
n-Undecane for synthesis, Sigma-Aldrich®
n-Undecane
Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196
Melting Pt: -26.7
Flash Pt: 60
MDL Number: MFCD00008959
CAS Number: 1120-21-4
UN: 2330
ADR: 3 III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -26 °C
Molar Mass 156.31 g/mol
Boiling Point 195 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 60 °C
Explosion Limit 0.6 - 6.5 %(V)
Density 0.74 g/cm3 (20 °C)
Ignition Point 220 °C
Viscosity Kinematic  - 7 mm2/s (40 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR