(±)-Linalool for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18627.0005 8.18627.0250 8.18627.0005 8.18627.0250
EM8.18627.0005CS 5942.38 CAD
EM8.18627.0005 EM8.18627.0250 CA8.18627.0005 CA8.18627.0250
(±)-Linalool for synthesis, Sigma-Aldrich®
(±)-Linalool
Formula: C₁₀H₁₈O
MDL Number: MFCD00008906
CAS Number: 78-70-6

Order Now

Specification Test Results

Solubility 1.45 g/l (20 °C)
Molar Mass 154.25 g/mol
Boiling Point 198 - 200 °C
Vapor Pressure 0.10 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.462 (20 °C, 589 nm)
Explosion Limit 0.9 - 5.2 %(V)
Density 0.86 g/cm3 (20 °C)
pH 4.5 (1.45 g/l, H2O, 25 °C)
Ignition Point 235 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR