Triisobutyl phosphate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18606.0100 8.18606.0500 8.18606.1000 8.18606.0100 8.18606.0500 8.18606.1000
EM8.18606.0100CS 1094.94 CAD
EM8.18606.0100 EM8.18606.0500 EM8.18606.1000 CA8.18606.0100 CA8.18606.0500 CA8.18606.1000
Triisobutyl phosphate for synthesis, Sigma-Aldrich®
Triisobutyl phosphate
Formula: C₁₂H₂₇O₄P
CAS Number: 126-71-6

Order Now

Specification Test Results

Solubility 0.26 g/l (20 °C)
Molar Mass 266.32 g/mol
Boiling Point 272 °C (1013 hPa) decomposes
Vapor Pressure 2 hPa (103 °C)
Flash Point 133 °C DIN 51758
Density 0.965 g/cm3 (20 °C)
Ignition Point 430 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR