Disodium succinate, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18601.0100 8.18601.0500 8.18601.5000 8.18601.0100 8.18601.0500 8.18601.5000
EM8.18601.0100CS 1022.26 CAD
EM8.18601.0100 EM8.18601.0500 EM8.18601.5000 CA8.18601.0100 CA8.18601.0500 CA8.18601.5000
Disodium succinate, anhydrous for synthesis, Sigma-Aldrich®
Disodium succinate
Formula: C₄H₄Na₂O₄
MDL Number: MFCD00002790
CAS Number: 150-90-3

Order Now

Specification Test Results

Solubility 350 g/l (25 °C)
Melting Point 300 °C
Molar Mass 162.05 g/mol
pH 7 - 9 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR