2,3-Pentanedione for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18555.0025 8.18555.0100 8.18555.0025 8.18555.0100
EM8.18555.0025CS 4472.98 CAD
EM8.18555.0025 EM8.18555.0100 CA8.18555.0025 CA8.18555.0100
2,3-Pentanedione for synthesis, Sigma-Aldrich®
2,3-Pentanedione
Formula: C₅H₈O₂
MDL Number: MFCD00009313
CAS Number: 600-14-6
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Solubility 60 g/l (15 °C)
Melting Point -52 °C
Molar Mass 100.12 g/mol
Boiling Point 110 - 112 °C
Vapor Pressure 28.5 hPa (20 °C)
Flash Point 18 °C
Refractive Index 1.4013 (19 °C, 589 nm)
Explosion Limit 1.8 - 10.9 %(V)
Density 0.96 g/cm3 (20 °C)
pH 4 (H2O)
Ignition Point 265 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR