2-(2-Aminoethoxy)ethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18431.0100 8.18431.0500 8.18431.0100 8.18431.0500
EM8.18431.0100CS 1306.66 CAD
EM8.18431.0100 EM8.18431.0500 CA8.18431.0100 CA8.18431.0500
2-(2-Aminoethoxy)ethanol for synthesis, Sigma-Aldrich®
2-(2-Aminoethoxy)ethanol
Formula: C₄H₁₁NO₂
MDL Number: MFCD00008181
CAS Number: 929-06-6
UN: 3055
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -11 °C
Molar Mass 105.14 g/mol
Boiling Point 125 - 131 °C (30 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 127 °C
Explosion Limit 2.0 - 15.5 %(V)
Density 1.06 g/cm3 (20 °C)
pH 10.2 (10 g/l, H2O, 20 °C)
Ignition Point 370 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR