Cetyldimethylethylammonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide

M-Clarity™ quality level = MQ200

8.18424.0100 8.18424.9025 8.18424.0100 8.18424.9025
EM8.18424.0100CS 2142.48 CAD
EM8.18424.0100 EM8.18424.9025 CA8.18424.0100 CA8.18424.9025
Cetyldimethylethylammonium bromide for synthesis, Sigma-Aldrich®
Cetyldimethylethylammonium bromide
Formula: C₂₀H₄₄BrN
MW: 378.4741 g/mol
Melting Pt: 178…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00011774
CAS Number: 124-03-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 200 g/l (20 °C)
Melting Point 178 - 186 °C
Molar Mass 378.47 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR