3,4-Diaminopyridine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine

M-Clarity™ quality level = MQ200

8.18392.0001 8.18392.0005 8.18392.0001 8.18392.0005
EM8.18392.0001CS 12093.32 CAD
EM8.18392.0001 EM8.18392.0005 CA8.18392.0001 CA8.18392.0005
3,4-Diaminopyridine for synthesis, Sigma-Aldrich®
3,4-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD00006401
CAS Number: 54-96-6
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Solubility 24 g/l (20 °C)
Melting Point 217 - 220 °C
Molar Mass 109.13 g/mol
Flash Point 240 °C
Ignition Point 480 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR