Hydrogen peroxide-urea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18356.0100 8.18356.0500 8.18356.0100 8.18356.0500
EM8.18356.0100CS 1251.36 CAD
EM8.18356.0100 EM8.18356.0500 CA8.18356.0100 CA8.18356.0500
Hydrogen peroxide-urea for synthesis, Sigma-Aldrich®
Hydrogen peroxide-urea
Formula: CH₆N₂O₃
MDL Number: MFCD00013119
CAS Number: 124-43-6
UN: 1511
ADR: 5.1 (8),III

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 75 - 85 °C (decomposition)
Molar Mass 94.07 g/mol
pH 6.3 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR