5-Bromovaleric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18329.0050 8.18329.0250 8.18329.0050 8.18329.0250
EM8.18329.0050CS 32025.02 CAD
EM8.18329.0050 EM8.18329.0250 CA8.18329.0050 CA8.18329.0250
5-Bromovaleric acid for synthesis, Sigma-Aldrich®
5-Bromovaleric acid
Formula: BrH₂C(CH₂)₃COOH
MDL Number: MFCD00004414
CAS Number: 2067-33-6
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 38 - 41 °C
Molar Mass 181.02 g/mol
Boiling Point 142 - 145 °C (17 hPa)
Flash Point 112 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR