1-(Trimethylsilyl)imidazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: TSIM, N-(Trimethylsilyl)imidazole, N-(Trimethylsilyl)-1H-imidazole, TMSI, 1-(Trimethylsilyl)-1H-imidazole

M-Clarity™ quality level = MQ200

8.18204.0025 8.18204.0100 8.18204.0025 8.18204.0100
EM8.18204.0025CS 10718.72 CAD
EM8.18204.0025 EM8.18204.0100 CA8.18204.0025 CA8.18204.0100
1-(Trimethylsilyl)imidazole for synthesis, Sigma-Aldrich®
1-(Trimethylsilyl)imidazole
Formula: C₆H₁₂N₂Si
MW: 140.26 g/mol
MDL Number: MFCD00005280
CAS Number: 18156-74-6

Order Now

Specification Test Results

Solubility (20 °C) Risk of violent reaction.
Melting Point -42 °C
Molar Mass 140.25 g/mol
Boiling Point 212 °C (decomposition)
Density 0.95 g/cm3 (25 °C)
Ignition Point 540 °C DIN 51794
Viscosity Kinematic 2.5 mm2/s (25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR