Sodium methoxide 30% in methanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18194.0100 8.18194.1000 8.18194.2500 8.18194.9050 8.18194.0100 8.18194.1000 8.18194.2500 8.18194.9050
EM8.18194.0100CS 850.04 CAD
EM8.18194.0100 EM8.18194.1000 EM8.18194.2500 EM8.18194.9050 CA8.18194.0100 CA8.18194.1000 CA8.18194.2500 CA8.18194.9050
Sodium methoxide 30% in methanol for synthesis, Sigma-Aldrich®
Sodium methoxide
Formula: CH₃NaO
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: 2…6 °C
Density: 0.97 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1289
ADR: 3.8,III

Order Now

Specification Test Results

Solubility (20 °C) (reaction)
Melting Point 2 - 6 °C
Boiling Point 92 °C (1013 hPa)
Vapor Pressure 34 hPa (20 °C)
Flash Point 33 °C
Explosion Limit 5.5 - 36.5 %(V) Methanol
Density 0.97 g/cm3 (20 °C)
pH 11 (20 g/l, H2O, 20 °C)
Ignition Point 455 °C Methanol

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR