2,2,6,6-Tetramethylpiperidine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.18132.0025
8.18132.0100
8.18132.0025
8.18132.0100
EM8.18132.0025CS
13858.18
CAD
EM8.18132.0025
EM8.18132.0100
CA8.18132.0025
CA8.18132.0100
2,2,6,6-Tetramethylpiperidine for synthesis, Sigma-Aldrich®
2,2,6,6-Tetramethylpiperidine
Formula:
C₉H₁₉N MW: 141.26 g/mol Boiling Pt: 152 Flash Pt: 24 |
MDL Number:
MFCD00005985 CAS Number: 768-66-1 UN: 2734 ADR: 8 (3) II |
Specification Test Results
Solubility | 25.9 g/l (20 °C) |
Melting Point | -59 °C |
Molar Mass | 141.25 g/mol |
Boiling Point | 152 - 156 °C |
Vapor Pressure | 14 hPa (39 °C) |
Flash Point | 37 °C |
Density | 0.83 g/cm3 (20 °C) |
Ignition Point | 290 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...