Methyltrialkylammonium chloride (mixture of C8-C10) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18079.0005 8.18079.0250 8.18079.1000 8.18079.0005 8.18079.0250 8.18079.1000
EM8.18079.0005CS 64437.14 CAD
EM8.18079.0005 EM8.18079.0250 EM8.18079.1000 CA8.18079.0005 CA8.18079.0250 CA8.18079.1000
Methyltrialkylammonium chloride (mixture of C8-C10) for synthesis, Sigma-Aldrich®
Methyltrialkylammonium chloride (mixture of C8-C10)
Formula: CH₃N(C₈-C₁₀n-Alkyl)₃Cl
MDL Number: MFCD00011862
CAS Number: 63393-96-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Boiling Point 240 °C
Flash Point 113 °C
Density 0.89 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR