Palladium(II) acetate Pd 47% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Palladium diacetate, Palladium (II) acetate

M-Clarity™ quality level = MQ200

8.18056.0001 8.18056.0005 8.18056.8500 8.18056.0001 8.18056.0005 8.18056.8500
EM8.18056.0001CS 749648.38 CAD
EM8.18056.0001 EM8.18056.0005 EM8.18056.8500 CA8.18056.0001 CA8.18056.0005 CA8.18056.8500
Palladium(II) acetate Pd 47% for synthesis, Sigma-Aldrich®
Palladium (II) acetate
Formula: C₄H₆O₄Pd
MDL Number: MFCD00012453
CAS Number: 3375-31-3

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 205 °C
Molar Mass 224.5 g/mol
pH 2 - 3 (H2O, 20 °C) (aqueous suspension)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR