Cyclopropylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Aminocyclopropane

M-Clarity™ quality level = MQ200

8.18017.0010 8.18017.0100 8.18017.0010 8.18017.0100
EM8.18017.0010CS 58632.22 CAD
EM8.18017.0010 EM8.18017.0100 CA8.18017.0010 CA8.18017.0100
Cyclopropylamine for synthesis, Sigma-Aldrich®
Cyclopropylamine
Formula: C₃H₇N
MW: 57.1
Boiling Pt: 49
Melting Pt: -10
Density: 0.8131
Flash Pt: 1
MDL Number: MFCD00001301
CAS Number: 765-30-0
UN: 2733
ADR: 3 (8) II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -10 °C
Molar Mass 57.09 g/mol
Boiling Point 49 - 50 °C
Flash Point 1 °C
Refractive Index 1.4206 (20 °C, 589 nm)
Density 0.82 g/cm3 (20 °C)
Ignition Point 275 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR