Silver trifluoroacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Silver(I) trifluoroacetate

M-Clarity™ quality level = MQ200

8.18003.0010 8.18003.0025 8.18003.0010 8.18003.0025
EM8.18003.0010CS 40743.46 CAD
EM8.18003.0010 EM8.18003.0025 CA8.18003.0010 CA8.18003.0025
Silver trifluoroacetate for synthesis, Sigma-Aldrich®
Silver trifluoroacetate
Formula: C₂AgF₃O₂
MW: 220.88 g/mol
MDL Number: MFCD00013199
CAS Number: 2966-50-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 257 - 260 °C
Molar Mass 220.88 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR