2,2-Bis(hydroxymethyl)propionic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid

M-Clarity™ quality level = MQ200

8.14996.0250 8.14996.1000 8.14996.0250 8.14996.1000
EM8.14996.0250CS 788.42 CAD
EM8.14996.0250 EM8.14996.1000 CA8.14996.0250 CA8.14996.1000
2,2-Bis(hydroxymethyl)propionic acid for synthesis, Sigma-Aldrich®
2,2-Bis(hydroxymethyl)propionic acid
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
MDL Number: MFCD00004199
CAS Number: 4767-03-7

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 188 - 191 °C
Molar Mass 134.13 g/mol
Flash Point 150 °C
pH 2.6 (50 g/l, H2O, 20 °C)
Ignition Point 300 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR