Methyl octanoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate

M-Clarity™ quality level = MQ200

8.14927.0025 8.14927.0100 8.14927.0025 8.14927.0100
EM8.14927.0025CS 5909.2 CAD
EM8.14927.0025 EM8.14927.0100 CA8.14927.0025 CA8.14927.0100
Methyl octanoate for synthesis, Sigma-Aldrich®
Methyl octanoate
Formula: C₉H₁₈O₂
MDL Number: MFCD00009551
CAS Number: 111-11-5

Order Now

Specification Test Results

Solubility 0.064 g/l (20 °C)
Melting Point -40 °C
Molar Mass 158.24 g/mol
Boiling Point 193 °C (1013 hPa)
Vapor Pressure 1.33 hPa (34.2 °C)
Flash Point 68 °C
Density 0.877 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR